Skip to Content
Merck
All Photos(2)

Key Documents

A7413

Sigma-Aldrich

Alloxan monohydrate

98%

Synonym(s):

2,4,5,6(1H,3H)-Pyrimidinetetrone, 2,4,5,6-Tetraoxypyrimidine, 5,6-Dioxyuracil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2N2O4 · H2O
CAS Number:
Molecular Weight:
160.08
Beilstein:
4573597
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder or crystals

mp

255 °C (dec.) (lit.)

solubility

H2O: may be hazy yellow

storage temp.

2-8°C

SMILES string

[H]O[H].O=C1NC(=O)C(=O)C(=O)N1

InChI

1S/C4H2N2O4.H2O/c7-1-2(8)5-4(10)6-3(1)9;/h(H2,5,6,8,9,10);1H2

InChI key

DSXMTJRUNLATRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Alloxan monohydrate can be used as a reactant
  • To produce the purple dye murexide.
  • In the synthesis of flavins via condensation with 1,2-diamines.
It can also be used to catalyze the formation of superoxide free radicals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ailton Santos Sena Júnior et al.
BMC sports science, medicine & rehabilitation, 12, 45-45 (2020-08-11)
Type 1 diabetes mellitus (T1DM) is a metabolic disease characterized by hyperglycemia and excessive generation of reactive oxygen species caused by autoimmune destruction of beta-cells in the pancreas. Among the antioxidant compounds, Curcuma longa (CL) has potential antioxidant effects and
Siamak Kazemi-Darabadi et al.
International journal of surgery (London, England), 12(8), 751-756 (2014-06-28)
Healing of skin wound is a multi-factorial and complex process. Treatment of diabetic wounds is still a major clinical challenge. Recently, stem cell transplantation to chronic wounds is favored. The objective of this study was to evaluate effects of pre-labeled
Leandro P Moura et al.
Lipids in health and disease, 10, 77-77 (2011-05-17)
Diabetes mellitus is associated with metabolic dysfunctions, including alterations in circulating lipid levels and fat tissue accumulation, which causes, among other pathologies, non-alcoholic fatty liver disease (NAFLD). The objective of this study was to analyse the effects of physical exercise
Krzysztof Krawczyk et al.
Science (New York, N.Y.), 368(6494), 993-1001 (2020-05-30)
Sophisticated devices for remote-controlled medical interventions require an electrogenetic interface that uses digital electronic input to directly program cellular behavior. We present a cofactor-free bioelectronic interface that directly links wireless-powered electrical stimulation of human cells to either synthetic promoter-driven transgene
Mohamed A Amin et al.
Archives of pharmacal research, 37(8), 1016-1031 (2013-12-03)
Diabetes is one of the leading causes of impaired wound healing. The objective of this study was to develop a bee venom-loaded wound dressing with an enhanced healing and anti-inflammatory effects to be examined in diabetic rats. Different preparations of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service