Skip to Content
Merck
All Photos(3)

Key Documents

A7205

Sigma-Aldrich

3-Acetamidophenol

97%

Synonym(s):

3′-Hydroxyacetanilide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$289.00

$289.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$289.00

About This Item

Linear Formula:
CH3CONHC6H4OH
CAS Number:
Molecular Weight:
151.16
Beilstein:
907998
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$289.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

crystals

mp

145-148 °C (lit.)

SMILES string

CC(=O)Nc1cccc(O)c1

InChI

1S/C8H9NO2/c1-6(10)9-7-3-2-4-8(11)5-7/h2-5,11H,1H3,(H,9,10)

InChI key

QLNWXBAGRTUKKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M S Rashed et al.
Chemical research in toxicology, 2(1), 41-45 (1989-01-01)
3'-Hydroxyacetanilide (AMAP) is a nonhepatotoxic regioisomer of acetaminophen (APAP) that nonetheless does form reactive metabolites which bind to hepatic proteins. Because differences in the nature of reactive metabolites formed from AMAP and APAP may explain differences in their propensity to
Robert H Pierce et al.
Biochemical pharmacology, 64(3), 413-424 (2002-07-31)
Overdose of the popular, and relatively safe, analgesic acetaminophen (N-acetyl-p-aminophenol, APAP, paracetamol) can produce a fatal centrilobular liver injury. APAP-induced cell death was investigated in a differentiated, transforming growth factor alpha (TGFalpha)-overexpressing, hepatocyte cell line and found to occur at
Metabolic alterations resulting from the inhibition of mitochondrial respiration by acetaminophen in vivo.
R L Esterline et al.
Biochemical pharmacology, 38(14), 2390-2392 (1989-07-15)
T G Myers et al.
Chemical research in toxicology, 8(3), 403-413 (1995-04-01)
Acetaminophen (4'-hydroxyacetanilide), a widely used analgesic/antipyretic drug, is hepatotoxic in large doses, whereas the m-hydroxy isomer of acetaminophen, 3'-hydroxyacetanilide, is not hepatotoxic. Both are oxidized by mouse liver cytochromes P-450 to reactive metabolites that bind covalently to hepatic proteins. Because
M A Bae et al.
Molecular pharmacology, 60(4), 847-856 (2001-09-20)
Acetaminophen (AAP), a widely used analgesic drug, can damage various organs when taken in large doses. In this study, we investigate whether AAP causes cell damage by altering the early signaling pathways associated with cell death and survival. AAP caused

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service