Skip to Content
Merck
All Photos(1)

Key Documents

A4571

Sigma-Aldrich

3-(α-Acetonylbenzyl)-4-hydroxycoumarin sodium salt

≥98%

Synonym(s):

4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$273.00

$273.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$273.00

About This Item

Linear Formula:
C19H15O4Na
CAS Number:
Molecular Weight:
330.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$273.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98%

form

powder

impurities

water, may contain traces
~8% Isopropanol

SMILES string

[Na].CC(=O)CC(c1ccccc1)C2=C(O)c3ccccc3OC2=O

InChI

1S/C19H16O4.Na/c1-12(20)11-15(13-7-3-2-4-8-13)17-18(21)14-9-5-6-10-16(14)23-19(17)22;/h2-10,15,21H,11H2,1H3;/q;+1/p-1

InChI key

KYITYFHKDODNCQ-UHFFFAOYSA-M

Gene Information

human ... VKORC1(79001)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 3 - Repr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tse-Yin Huang et al.
Scientific reports, 10(1), 15910-15910 (2020-09-29)
Resveratrol (RVT) has various beneficial bioactivities and popularly used as a dietary supplement. RVT showed inhibitions on CYP1A2/2C9/3A4, breast cancer resistance protein (BCRP), and some conjugated metabolites of RVT also inhibited BCRP. (±)Warfarin, an anticoagulant for cardiovascular disease but with
Wenjing Wang et al.
Journal of biomolecular structure & dynamics, 38(11), 3178-3187 (2019-08-06)
Empagliflozin is an oral sodium glucose co-transporter-2 inhibitor for type 2 diabetes mellitus. The interaction between empagliflozin and human serum albumin (HSA) was investigated experimentally and theoretically. Fluorescence quenching and time-resolved fluorescence spectroscopy indicated that the quenching mechanism of empagliflozin
Catherine K Chang et al.
Journal of vascular surgery, 60(6), 1554-1564 (2014-07-22)
Despite poor long-term patency, acceptable limb salvage has been reported with cryopreserved saphenous vein bypass (CVB) for various indications. However, utility of CVB in patients with critical limb ischemia (CLI) remains undefined. The purpose of this analysis was to determine
Akhtar Siddiqui et al.
International journal of pharmaceutics, 493(1-2), 1-6 (2015-07-26)
The objective of this study was to develop and validate XRPD analytical method for the estimation of percent crystalline warfarin sodium present in drug products. Warfarin sodium (WS) is a clathrate containing Isopropyl alcohol entrapped in the crystalline structure. Four
Frieder Schlunk et al.
Translational stroke research, 6(2), 133-139 (2014-11-27)
Little is known about the pathophysiology of oral anticoagulation-associated intracerebral hemorrhage (OAC-ICH). We compared hematoma volume, number of terminal deoxynucleotidyl dUTP nick-end labeling (TUNEL)-positive cells (indicating cell death), MMP-9 levels, and perilesional edema formation between warfarin-treated mice and controls. Intracerebral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service