Skip to Content
Merck
All Photos(1)

Key Documents

A1606

Sigma-Aldrich

4-Acetamidoantipyrine

97%

Synonym(s):

4-Acetylaminophenazone, N-(2,3-Dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)acetamide, N-Antipyrinylacetamide, NSC 331807

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15N3O2
CAS Number:
Molecular Weight:
245.28
Beilstein:
234585
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

200-203 °C (lit.)

SMILES string

CN1N(C(=O)C(NC(C)=O)=C1C)c2ccccc2

InChI

1S/C13H15N3O2/c1-9-12(14-10(2)17)13(18)16(15(9)3)11-7-5-4-6-8-11/h4-8H,1-3H3,(H,14,17)

InChI key

OIAGWXKSCXPNNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Heinemeyer et al.
European journal of clinical pharmacology, 45(5), 445-450 (1993-01-01)
We have studied the clearance of monomethylaminoantipyrine (MMAAP), the pharmacologically active form of metamizol, in 46 patients in surgical intensive care with different degrees of renal dysfunction. In 23 patients without any renal impairment, mean clearance was 2.8 ml.min-1 x
Further metabolism of 4-acetylaminoantipyrine, the major metabolite of aminopyrine, in rats.
T Tanaka et al.
Chemical & pharmaceutical bulletin, 35(8), 3519-3522 (1987-08-01)
E Neddermann et al.
European journal of drug metabolism and pharmacokinetics, 13(2), 105-111 (1988-04-01)
Metabolites of dipyrone have been determined in the saliva of 18 volunteers following the oral intake of 0.5 g, 1.0 g, 1.5 g, 2.0 g and 2.5 g dipyrone. High concentrations were measured for N-methyl-aminoantipyrine (MAA), the other analgetic active
S C Pierre et al.
British journal of pharmacology, 151(4), 494-503 (2007-04-17)
Dipyrone is a potent analgesic drug that has been demonstrated to inhibit cyclooxygenase (COX). In contrast to classical COX-inhibitors, such as aspirin-like drugs, dipyrone has no anti-inflammatory effect and a low gastrointestinal toxicity, indicating a different mode of action. Here
M Levy et al.
European journal of clinical pharmacology, 27(4), 453-458 (1984-01-01)
The pharmacokinetics of the dipyrone metabolites 4-methylaminoantipyrine (MAA), 4-aminoantipyrine (AA), 4-formylaminoantipyrine (FAA) and 4-acetylaminoantipyrine (AAA) were evaluated following the administration of a single oral 1.0 g dose of dipyrone to 23 healthy volunteers. Twelve were slow and 11 were rapid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service