Skip to Content
Merck
All Photos(1)

Key Documents

BASMF2012

Sigma-Aldrich

Glassy Carbon Voltammetry Electrode

Synonym(s):

Glassy Carbon Working Electrode for Voltammetry

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$625.00

$625.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$625.00

About This Item

UNSPSC Code:
12352300
NACRES:
NA.23

$625.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Application

Glassy carbon working electrode is the most commonly used carbon-based electrode in electrochemistry such as cyclic voltammetry and linear sweep voltammetry due to its properties of high temperature resistance, hardness, low density and low electrical resistance. Glassy carbon is one of the widely used electrode materials for making working electrodes. With a wide potential window and good electrical conductivity, glassy carbon is electrochemically inert, dense, and hard as glass, without gas or liquid permeability. In addition to very high thermal stability, glassy carbon is extremely resistant to chemical attack.

Legal Information

Product of Bioanalytical Systems, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. hello, can you provide diameter of this product please

    1 answer
    1. As listed in the PROPERTIES section on the product page, the diameter is 3.0mm.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service