Skip to Content
Merck
All Photos(1)

Key Documents

94908

Sigma-Aldrich

Vinylbenzyl chloride

purum, mixture of isomers, ≥95.0%, stabilized

Synonym(s):

(Chloromethyl)styrene

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$214.00

$214.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$214.00

About This Item

Linear Formula:
H2C=CHC6H4CH2Cl
CAS Number:
Molecular Weight:
152.62
Beilstein:
2204384
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$214.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

5.3 (vs air)

vapor pressure

1 mmHg ( 56.1 °C)

grade

purum

Assay

≥95.0%

form

liquid

autoignition temp.

1139 °F

refractive index

n20/D 1.572 (lit.)
n20/D 1.572

bp

229 °C (lit.)

density

1.074 g/mL at 25 °C (lit.)

shipped in

wet ice

storage temp.

−20°C

SMILES string

ClCc1ccc(C=C)cc1.ClCc2cccc(C=C)c2

InChI

1S/2C9H9Cl/c1-2-8-3-5-9(7-10)6-4-8;1-2-8-4-3-5-9(6-8)7-10/h2*2-6H,1,7H2

InChI key

QSDACPJNNYTTDP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • RAFT homopolymerization of vinylbenzyl chloride with benzyl ethyl trithiocarbonate and synthesis of block copolymers from poly (VBC) macro-RAFT agent: The study discusses living radical polymerizations applied to the polymerization of vinylbenzyl chloride (Sato et al., 2014).

  • Quaternized poly (styrene-co-vinylbenzyl chloride) anion exchange membranes for alkaline water electrolysers: This paper details the use of a styrene and vinylbenzyl chloride co-polymer as the polymer backbone for developing anion exchange membranes (Vengatesan et al., 2015).

  • Polybenzimidazole-crosslinked poly (vinylbenzyl chloride) with quaternary 1, 4-diazabicyclo (2.2. 2) octane groups as high-performance anion exchange membrane: The research highlights the crosslinking of polybenzimidazole with poly(vinylbenzyl chloride), enhancing the membrane′s flexibility and strength in various states (Lu et al., 2015).

  • Preparation and characterization of sulfur-vinylbenzyl chloride polymer under optimized reaction conditions using inverse vulcanization: This study focuses on the reaction of 4-vinylbenzyl chloride with sulfur under optimized conditions to produce a linear sulfur-based polymer (Abbasi et al., 2021).

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Guerritore et al.
Polymers, 12(6) (2020-06-25)
The development of new styrene-based hyper-crosslinked nanocomposites (HCLN) containing mesoporous silica nanoparticles (MSN) is reported here as a new strategy to obtain functional high surface area materials with an enhanced hydrophilic character. The HCLN composition, morphology and porous structure were
Véronique Lenoble et al.
Journal of colloid and interface science, 448, 473-481 (2015-03-17)
Three novel Ni(II)-Ion-Imprinted Polymer (IIP) were synthesized by precipitation polymerization of ethylene glycol dimethacrylate (crosslinker) with a complex of nickel(II) and vinylbenzyl iminodiacetic acid (VbIDA). The three IIPs were prepared with various mixtures of porogen solvents: methanol, methanol/2-methoxyethanol and methanol/acetonitrile

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service