Skip to Content
Merck
All Photos(1)

Key Documents

94200

Sigma-Aldrich

Undecylamine

≥98.0% (GC)

Synonym(s):

1-Amino-undecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10NH2
CAS Number:
Molecular Weight:
171.32
Beilstein:
1736378
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
≥98.0% (GC)

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.439 (lit.)
n20/D 1.439

bp

106-107 °C/8 mmHg (lit.)
240 °C (lit.)

mp

15-17 °C (lit.)

density

0.796 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCCCCCCCCCCN

InChI

1S/C11H25N/c1-2-3-4-5-6-7-8-9-10-11-12/h2-12H2,1H3

InChI key

QFKMMXYLAPZKIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mara Canovi et al.
Sensors (Basel, Switzerland), 12(12), 16420-16432 (2013-02-28)
Great interest is currently being devoted to the development of nanoparticles (NPs) for biomedical purposes, designed to improve the pharmacokinetic profile of their cargos (either imaging probes or drugs) and to enhance the specific targeting at the disease site. Recent
Hideko Sahara et al.
Journal of oleo science, 65(10), 861-865 (2016-09-16)
The degree of counterion binding (β) to micelles of aqueous n-alkylammonium chlorides (C
Qixuan Zhong et al.
Nano letters, 19(6), 4151-4157 (2019-05-24)
X-type ligands, for example, the pair of oleylamine (OAm) and oleic acid (OA), have been widely used to prepare CsPbX3 nanocrystals (NCs). However, the proton exchange between coordinated OAm and OA may induce the detachment of ligands, resulting in poor
Asja Pettignano et al.
Polymers, 11(7) (2019-07-26)
The present work explores the possibility of chemically modifying carboxymethyl cellulose (CMC), a widely diffused commercial cellulose ether, by grafting of hydrophobic moieties. Amidation of CMC, at high temperature and in heterogeneous conditions, was selected as synthetic tool for grafting
Jonathan O'Keeffe Ahern et al.
Biomacromolecules, 19(5), 1410-1415 (2017-11-11)
Using a combined synthesis approach comprising reversible addition-fragmentation transfer polymerization and ring opening reaction, a series of poly glycidyl methacrylate (polyGMA) polymers were designed and synthesized for gene delivery. These polymers characterized by low cationic charge respective to established gene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service