Skip to Content
Merck
All Photos(1)

Key Documents

936383

Sigma-Aldrich

Mc-Val-Cit-PAB

≥95%

Synonym(s):

6-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)-N-((S)-1-(((S)-1-((4-(hydroxymethyl)phenyl)amino]-1-oxo-5-ureidopentan-2-yl)amino]-3-methyl-1-oxobutan-2-yl)hexanamide, N-[(1S)-1-[[(1S)-4-(carbamoylamino)-1-[[4-(hydroxymethyl)phenyl]carbamoyl]butyl]carbamoyl]-2-methylpropyl]-6-(2,5-dioxopyrrol-1-yl)hexanamide, N-[6-(2,5-Dihydro-2,5-dioxo-1H-pyrrol-1-yl)-1-oxohexyl]-L-valyl-N5-(aminocarbonyl)-N-[4-(hydroxymethyl)phenyl]-L-ornithinamide (ACI), L-Ornithinamide, N-[6-(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)-1-oxohexyl]-L-valyl-N5-(aminocarbonyl)-N-[4-(hydroxymethyl)phenyl]- (9CI, ACI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H40N6O7
Molecular Weight:
572.65
MDL number:
UNSPSC Code:
12352209
Pricing and availability is not currently available.

Quality Level

Assay

≥95%

form

powder or crystals

color

white to yellow

storage temp.

−20°C

SMILES string

[C@@H](NC([C@@H](NC(CCCCCN1C(=O)C=CC1=O)=O)[C@H](C)C)=O)(C(NC2=CC=C(CO)C=C2)=O)CCCNC(N)=O

InChI

1S/C28H40N6O7/c1-18(2)25(33-22(36)8-4-3-5-16-34-23(37)13-14-24(34)38)27(40)32-21(7-6-15-30-28(29)41)26(39)31-20-11-9-19(17-35)10-12-20/h9-14,18,21,25,35H,3-8,15-17H2,1-2H3,(H,31,39)(H,32,40)(H,33,36)(H3,29,30,41)/t21-,25-/m0/s1

InChI key

AMKBTTRWLGVRER-OFVILXPXSA-N

Application

This is a bifunctional cathepsin cleavable linker used in the construction of antibody drug conjugates (ADC).

Features and Benefits

The maleimide functionality can be used for bioconjugation to thiols in monoclonal antibodies while the benzyl alcohol can be used for construction of carbamate linkages with amine containing payloads.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qinhuai Lai et al.
European journal of medicinal chemistry, 199, 112364-112364 (2020-05-14)
Cryptophycin-52 (CR52), a tubulin inhibitor, exhibits promising antitumor activity in vitro (picomolar level) and in mouse xenograft models. However, the narrow therapeutic window in clinical trials limits its further development. Antibody-drug conjugate (ADC), formed by coupling cytotoxic compound (payload) to an
Zixuan Ye et al.
International journal of nanomedicine, 16, 2443-2459 (2021-04-06)
Specific modifications to carriers to achieve targeted delivery of chemotherapeutics into malignant tissues are a critical point for efficient diagnosis and therapy. In this case, bovine serum albumin (BSA) was conjugated with cetuximab-valine-citrulline (vc)-doxorubicin (DOX) to target epidermal growth factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service