Skip to Content
Merck
All Photos(2)

Documents

935662

Sigma-Aldrich

Zinc sulfate heptahydrate

greener alternative

99.999% trace metals basis

Synonym(s):

White vitriol, Zinc vitriol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSO4 · 7H2O
CAS Number:
Molecular Weight:
287.56
MDL number:
UNSPSC Code:
51241886
NACRES:
NA.21

Quality Level

Assay

99.999% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤15.0 ppm (trace metals analysis)

pH

4-6 (20 °C, 50 g/L in water)

mp

100 °C

solubility

ethanol: sparingly soluble
glycerol: sparingly soluble
water: soluble

greener alternative category

SMILES string

O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.7H2O.Zn/c1-5(2,3)4;;;;;;;;/h(H2,1,2,3,4);7*1H2;/q;;;;;;;;+2/p-2

InChI key

RZLVQBNCHSJZPX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product has been enhanced for energy efficency. Click here for more information.
Zinc sulfate heptahydrate is a colorless solid and known as white vitriol. It is orthorhombic crystal, and at high temperature, it decomposes into zinc oxide and sulfur trioxide. Zinc sulfate heptahydrate is soluble in water, slightly soluble in ethanol and glycerol. Zinc sulfate is produced by dissolving Zinc metal in aqueous sulfuric acid. High-purity zinc oxide is treated with sulfuric acid to produce pharmaceutical-grade zinc sulphate.

Application

Zinc Sulfate Heptahydrate is widely used in the synthesis of ZnO nanoparticles. It has also been used as a template for solvothermal synthesis of nanoporous TiO2 spheres with tunable chamber structures ranging from hollow to solid and hybrid compositions. Lithium sulfate and Zinc sulfate are used as an additive to increase the life cycle, cathode stability and efficiency of lead acid batteries. In addition, Zinc sulfate is also used in multiple industrial applications such as animal feed as a supplement on growth performance. It is also used as an inhibitor of fungal growth[5]. In order to increase the thickness of the Zn plating as the zinc concentration increases, zinc sulphate heptahydrate is frequently used as an electrolyte in the electroplating industry[6].

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of Lithium Sulfate and Zinc Sulfate Additives on the Cycle Life and Efficiency of Lead Acid Batteries
Musei, N.N.
ACS Omega, 6 (2021)
Controlled Precipitation of Zinc Oxide Particles at Room Temperature
Oliveira, A. P. A.
Chemistry of Materials, 15 (2003)
Intelligent Hydrated-Sulfate Template Assisted Preparation of Nanoporous TiO2 Spheres and Their Visible-Light Application
Lu, X.
ACS Applied Materials & Interfaces, 3 (2011)
Effects of supplemental zinc sulfate on growth performance, carcass characteristics, and antimicrobial resistance in feedlot heifers
Drouillard, J.S.
Journal of Animal Science, 97 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service