Skip to Content
Merck
All Photos(1)

Key Documents

929557

Sigma-Aldrich

Thiamine-(4-methyl-13C-Thiazol-5-yl-13C3) monophosphate hydrochloride

≥99 atom % 13C, ≥98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C4C8H19Cl2N4O4PS
Molecular Weight:
421.22
UNSPSC Code:
12352116
NACRES:
NA.13
Pricing and availability is not currently available.

isotopic purity

≥99 atom % 13C

Quality Level

Assay

≥98% (CP)

technique(s)

mass spectrometry (MS): suitable

mass shift

M+4

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service