Skip to Content
Merck
All Photos(1)

Documents

929514

Sigma-Aldrich

ItHept HCl

≥95%

Synonym(s):

1,3-Bis(3-ethylpentan-3-yl)-1H-imidazol-3-ium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H33ClN2
Molecular Weight:
300.91
UNSPSC Code:
12352005
NACRES:
NA.21

Quality Level

Assay

≥95%

form

powder

SMILES string

CCC(CC)(CC)N1C=[N+](C(CC)(CC)CC)C=C1.[Cl-]

Application

ItHept HCl is an N-alkyl NHC ligand with demonstrated uses in the following reactions. Au-catalyzed hydration, Cu-catalyzed hydroboration, Ag-catalyzed hydroboration, Cu-catalyzed C–O coupling and Pd-catalyzed C–C and C–N couplings. This ligand features significantly higher steric volume compared with ItBu (659983) while maintaining the σ-donation and σ-acceptance inherent to N-alkyl NHC ligands.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The Hazari group has developed an improved palladium precatalyst scaffold for a wide range of cross-coupling reactions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service