Skip to Content
Merck
All Photos(1)

Key Documents

928623

Sigma-Aldrich

Chamfered tips

33G, 0.25", 25/box, black

Synonym(s):

3D bioprinting consumables, Bioprinting nozzle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
42142600
NACRES:
NA.25
Pricing and availability is not currently available.

description

Chamfered tips, 33G, 0.25", 25/box Black

Looking for similar products? Visit Product Comparison Guide

Application

Chamfered tips are ideal for dispensing between delicate surfaces or making exact, repeatable micro-deposits. The tips are designed with a special outer diameter taper that prevents wicking for greater deposit accuracy.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service