Skip to Content
Merck
All Photos(1)

Key Documents

926183

Sigma-Aldrich

Ru-MACHO®-BH

Takasago

Synonym(s):

(OC-6-13)-Carbonyl[2-(diphenylphosphino-κP)-N-[2-(diphenylphosphino-κP)ethyl]ethanamine-κN][tetrahydroborato(1-)-κH]ruthenium

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$60.80
1 G
$188.00
5 G
$559.00

$60.80


Estimated to ship onApril 11, 2025



Select a Size

Change View
250 MG
$60.80
1 G
$188.00
5 G
$559.00

About This Item

Empirical Formula (Hill Notation):
C29H34BNOP2Ru
CAS Number:
Molecular Weight:
586.41
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

$60.80


Estimated to ship onApril 11, 2025


Quality Level

form

powder

Application

Ru-MACHO®-BH novel ruthenium catalyst for homogeneous hydrogenations of esters under base free conditions.

Legal Information

Product of Takasago. For research and development purposes only. PCT/JP2010-004301
Ru-MACHO is a registered trademark of Takasago Intl. Corp.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic Hydrogenation of Esters. Development of an Efficient Catalyst and Processes for Synthesising (R)-1,2-Propanediol and 2-(l-Menthoxy)ethanol
Kuriyama W., et al.
Organic Process Research & Development, 16, 166?171-166?171 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service