Skip to Content
Merck
All Photos(1)

Key Documents

925918

Sigma-Aldrich

9-(2,6-Dimethylphenyl)-1-methoxy-10-phenylacridinium bromide

95%

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$256.00

$256.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$256.00

About This Item

Empirical Formula (Hill Notation):
C28H24BrNO
CAS Number:
Molecular Weight:
470.40
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.21

$256.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

190-198 (decomp)

SMILES string

COC1=CC=CC2=[N+](C3=CC=CC=C3)C4=CC=CC=C4C(C5=C(C)C=CC=C5C)=C21.[Br-]

InChI

1S/C28H24NO.BrH/c1-19-11-9-12-20(2)26(19)27-22-15-7-8-16-23(22)29(21-13-5-4-6-14-21)24-17-10-18-25(30-3)28(24)27;/h4-18H,1-3H3;1H/q+1;/p-1

InChI key

AIMWDQBPPADYLB-UHFFFAOYSA-M

Application

9-(2,6-Dimethylphenyl)-1-methoxy-10-phenylacridinium bromide is an acridinium photocatalyst used for a variety of transformations.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Legal Information

Product of Solvias

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bouthayna Zilate et al.
Chemical communications (Cambridge, England), 56(12), 1767-1775 (2020-01-31)
Recent developments in preparative photocatalysis have reshaped synthetic strategies and now represent an integral part of current organic chemistry. Due to their favourable electrochemical and photophysical properties, the nowadays most frequently used photocatalysts are based on precious Ru- and Ir-polypyridyl
Christian Fischer et al.
Angewandte Chemie (International ed. in English), 57(9), 2436-2440 (2017-12-19)
Despite the manifold use of heterocyclic fluorophores, only a fraction of the desired dye diversity can be accessed by contemporary synthetic approaches. Herein, we describe a modular method that converts various carboxylic acid esters directly into a broad spectrum of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service