Skip to Content
Merck
All Photos(1)

Key Documents

925497

Sigma-Aldrich

[Ir(dFOMeppy)2(dtbbpy)]PF6

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$186.00
500 MG
$1,200.00

$186.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$186.00
500 MG
$1,200.00

About This Item

Empirical Formula (Hill Notation):
C42H40F10IrN4O2P
CAS Number:
Molecular Weight:
1045.97
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

$186.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

mp

>300 °C

Application

[Ir(dFOMeppy)2(dtbbpy)]PF6 is a versatile photocatalyst that can be used for a variety of synthetic transformations.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chi Chip Le et al.
Journal of the American Chemical Society, 137(37), 11938-11941 (2015-09-04)
In this study we demonstrate that molecular fragments, which can be readily coupled via a simple, in situ RO-C═OR bond-forming reaction, can subsequently undergo metal insertion-decarboxylation-recombination to generate Csp(2)-Csp(3) bonds when subjected to metallaphotoredox catalysis. In this embodiment the conversion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service