Skip to Content
Merck
All Photos(1)

Key Documents

924385

Sigma-Aldrich

Silicon Tetrabiphenyl MeOTAD

greener alternative

Synonym(s):

Si-TetraBiphenyl MeOTAD, SiTBP-MeOTAD

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$495.00

$495.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$495.00

About This Item

Empirical Formula (Hill Notation):
C104H88N4O8Si
CAS Number:
Molecular Weight:
1549.92
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$495.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

Band Gap: 3.04 eV

Quality Level

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

white to off-white

Orbital energy

HOMO -5.34 eV 
LUMO -2.30 eV 

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Silicon Tetrabiphenyl MeOTAD is an organic molecule that is used as a hole transport material(HTM). Its properties include high solubility, good stability and an amorphous structue. Click here for more information.

Application

SiTP-OMeTPA is a hole transport material (HTM) that has been used in perovskite solar cells.[1] This material has a reversible redox peak in the cyclic a perovskite solar cell was Voc (0.99 V), Jsc (19.85 mA*cm-2), FF (71.69 %), PCE (14.15 %).[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular design with silicon core: toward commercially available hole transport materials for high-performance planar p?i?n perovskite solar cells.
Xue R, et al.
Journal of Material Chemistry A, 6, 404-413 (2018)

Articles

Novel biosensing devices, like bio-FET sensors, offer label-free analysis by detecting ionic and biomolecular charges, crucial for diagnostics and biological research.

Explore the eco-friendly potential of organic thin film transistors (OTFTs) for detecting chemical analytes, identifying viruses, and assisting in health diagnostics. This mini-review highlights challenges of achieving sustainability, safety, and biodegradability of each component of an OTFT sensor.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service