Skip to Content
Merck
All Photos(1)

Key Documents

922765

Sigma-Aldrich

IPr* HCl

Synonym(s):

N,N′-Bis(2,6-bis(diphenylmethyl)-4-methylphenyl)imidazolium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$142.00

$142.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$142.00

About This Item

Empirical Formula (Hill Notation):
C69H57N2Cl
CAS Number:
Molecular Weight:
949.66
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

$142.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

SMILES string

[Cl-].[n+]6(c[n](cc6)c7c(cc(cc7C(c%11ccccc%11)c%10ccccc%10)C)C(c9ccccc9)c8ccccc8)c1c(cc(cc1C(c5ccccc5)c4ccccc4)C)C(c3ccccc3)c2ccccc2

InChI key

SMTNUZDTTFSHEZ-UHFFFAOYSA-M

Application

IPr*HCl is a bulky, flexible NHC ligand used for organometallic catalysis.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guillaume Berthon-Gelloz et al.
Dalton transactions (Cambridge, England : 2003), 39(6), 1444-1446 (2010-01-28)
Herein, we wish to describe the efficient three-step synthesis of a novel highly hindered, but flexible, N-heterocyclic carbene and its coordination chemistry to Ag(i) and Rh(i).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service