Skip to Content
Merck
All Photos(1)

Key Documents

921033

Sigma-Aldrich

Palladium(II) chloride

≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$175.00
5 G
$691.00
25 G
$2,730.00

$175.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$175.00
5 G
$691.00
25 G
$2,730.00

About This Item

Empirical Formula (Hill Notation):
Cl2Pd
CAS Number:
Molecular Weight:
177.33
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.23
grade:
for analytical purposes

$175.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

≥99.99% trace metals basis

mp

678-680 °C (lit.)

density

4 g/mL at 25 °C (lit.)

SMILES string

Cl[Pd]Cl

InChI

1S/2ClH.Pd/h2*1H;/q;;+2/p-2

InChI key

PIBWKRNGBLPSSY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Application Guide for Palladium Catalyzed Cross-Coupling Reactions

Used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.
Palladium chloride is a transition metal catalyst that can be used to synthesize palladium complexes applicable in heterogeneous catalysis.[1][2] The Pd NPs synthesized from PdCl2 using oxytocin as a reductant showed remarkable catalytic activity in Suzuki reaction under green conditions.[3]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amidoxime modified PAN supported palladium complex: A greener and efficient heterogeneous catalyst for heck reaction.
Sruthi P R, et al.
Inorgorganica Chimica Acta, 502, 119305-119305 (2020)
Palladium supported on triazolyl-functionalized hypercrosslinked polymers as a recyclable catalyst for Suzuki?Miyaura coupling reactions
Liu C, et al.
Royal Society of Chemistry Advances, 10(29), 17123-17128 (2020)
A simple, efficient and green approach for the synthesis of palladium nanoparticles using Oxytocin: Application for ligand free Suzuki reaction and total synthesis of aspongpyrazine A.
Bendre A D,et al.
Journal of Organometallic Chemistry, 909, 121093-121093 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service