Skip to Content
Merck
All Photos(1)

Key Documents

920428

Sigma-Aldrich

Methoxypolyethylene glycol

average MN 1,000 (by NMR), glycol propyl amine, methoxy

Synonym(s):

PEG amine, Poly(ethylene) glycol amine, Polyethylene glycol, mPEG NH2, mPEG amine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$79.60

$79.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$79.60

About This Item

Linear Formula:
CH3(OCH2CH2)nOCH2CH2CH2NH2
UNSPSC Code:
12352116
NACRES:
NA.23

$79.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Methoxypolyethylene glycol propyl amine 1,000,

form

semisolid

Quality Level

mol wt

average Mn 1,000 (by NMR)

color

white to off-white

Ω-end

amine

α-end

methoxy

polymer architecture

shape: linear
functionality: monofunctional

Application

Methoxypolyethylene glycol propyl amine 1,000 is a pegylation agent used to conjugate proteins to reduce immunogenicity and prolong serum half-life of proteins and enzymes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Ton J Dirks et al.
Bioconjugate chemistry, 20(6), 1129-1138 (2009-05-21)
The Cu(I)-catalyzed azide-alkyne cycloaddition (CuAAC) has recently proven to be a powerful synthetic tool in various fields of chemistry, including protein-polymer conjugation. In this article, we describe a fluorogenic CuAAC, which allows for efficient monitoring of protein-polymer conjugation. We show

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service