Skip to Content
Merck
All Photos(2)

Key Documents

920304

Sigma-Aldrich

Indium Phosphide Zinc Sulfide (InP/ZnS) Quantum Dots

in toluene, stabilized with oleic acid, fluorescence λem 650 nm

Synonym(s):

Fluorescent nanocrystals, QDs

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$501.00

$501.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$501.00

About This Item

Linear Formula:
InP/ZnS
UNSPSC Code:
26111700
NACRES:
NA.23

$501.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

Quality Level

form

liquid

concentration

in toluene

fluorescence

FWHM 40-45 nm
λem 650 nm
, quantum yield ≥60%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Reactive group: Amine

Application

InP/ZnS core shell nanocrystals are Cadmium free/heavy metal free quantum dots suitable for different applications like light emitting diodes such as LEDs, displays, and biomedical applications. These Qdots are environmentally-friendly, heavy metal-free alternative to cadmium based quantum dots. These InP/ZnS core/shell quantum dots are non-toxic while offering high performance for various applications.

Caution

Do not freeze.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Lungs

WGK

WGK 3

Flash Point(F)

39.2 °F

Flash Point(C)

4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solar cell sensitized with ″green″ InP-ZnS quantum dots: Effect of ZnS shell deposition
Zavaraki J A, et al.
Nanostructured Conjugated Polymers for Photovoltaic Devices, 22 (2020)
Bing Chen et al.
Small (Weinheim an der Bergstrasse, Germany), 16(32), e2002454-e2002454 (2020-07-03)
InP quantum dots (QDs) are typical III-V group semiconductor nanocrystals that feature large excitonic Bohr radius and high carrier mobility. The merits of InP QDs include large absorption coefficient, broad color tunability, and low toxicity, which render them promising alternatives

Articles

Colloidal quantum dots are nano-meter sized, single-crystalline semiconductor fragments that can have highly customizable optical, electronic, and chemical properties through controlled synthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service