Skip to Content
Merck
All Photos(1)

Key Documents

920177

Sigma-Aldrich

Vanadium (V) oxide nanosheets

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$415.00

$415.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
$415.00

About This Item

Linear Formula:
V2O5
CAS Number:
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23

$415.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

SMILES string

[V](=O)(=O)(=O)(=O)(=O)[V]

InChI

1S/5O.2V

InChI key

GNTDGMZSJNCJKK-UHFFFAOYSA-N

Application

2D V2O5 (vanadium oxide) nanosheets have superior electrochemical performance in terms of their specific capacity, rate capability, and cyclability as cathode materials for LIBs. In particular, their unique nanoscale characteristics, including 2D morphology, hierarchical porous structure, large specific surface, opens up new opportunities in the development of high-performance next-generation LIBs used for alternative energy and electric transportation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nam-Soon Choi et al.
Angewandte Chemie (International ed. in English), 51(40), 9994-10024 (2012-09-12)
Energy-storage technologies, including electrical double-layer capacitors and rechargeable batteries, have attracted significant attention for applications in portable electronic devices, electric vehicles, bulk electricity storage at power stations, and "load leveling" of renewable sources, such as solar energy and wind power.
The Current Move of Lithium Ion Batteries Towards the Next Phase.
Kim T H, et al.
Advanced Energy Materials, 2, 860-860 (2012)
Building better batteries.
M Armand et al.
Nature, 451(7179), 652-657 (2008-02-08)

Questions

  1. are these nanosheets dispersed in a solvent? If this is a pure liquid what is the density?

    1 answer
    1. This product is a 0.1 - 0.5% solution of Vanadium (V) oxide nanosheets in water, containing 1 - 2% Tetrabutylammonium hydroxide as a dispersant. The density is not tested on a lot-to-lot basis, however, historical information shows a density of 0.992 g/mL. Please see the link below to access a sample or lot-specific Certificate of Analysis:
      https://www.sigmaaldrich.com/product/aldrich/920177#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service