Skip to Content
Merck
All Photos(1)

Key Documents

920061

Sigma-Aldrich

Graphene, monolayer film

small grain, CVD graphene on copper foil, A3 size

Synonym(s):

Roll-to-Roll graphene

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$3,900.00

$3,900.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$3,900.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12141908
NACRES:
NA.23

$3,900.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

R2R Monolayer small grain CVD graphene on copper foil, A3 size, avg. no. of layers, 1

Quality Level

description

Growth method: roll-to-roll CVD
Number of layer: Monolayer

Raman intensity 2D/G: ≥1.5

feature

avg. no. of layers 1

sheet resistance

240 Ω/sq ±10%

size

2.2 μm × 2.2 μm (±10%), grain size

thickness

35 μm , Cu Foil

surface coverage

surface coverage >98%

transmittance

>97%

semiconductor properties

(mobility>1500 cm2/V·s) (Hall effect measurements)

SMILES string

NC.NC.NC.NC.NC.NC.NC.NC.NC

InChI

1S/9CH5N/c9*1-2/h9*2H2,1H3

InChI key

DXIFQVRACSPGSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Roll-to-roll, high-quality, monolayer CVD graphene with small grain size (∼5μm2) on copper foil, A3 size.

Application

Our Roll-to-Roll CVD graphene products are true monolayer high quality graphene, fabricated inside a Cleanroom, heavily monitored and QC to assure high reproducibility.
The roll-to-roll process allows continuous, large scale graphene production.[1]

This small grain size product with low sheet resistance would enable unmatched reproducibility and allow high performance for CVD graphene based cell culture scaffold[2], biosensors[3] and chemically gated sensors.

Caution

Be cautious not to drop
Keep away from contamination, heat, dust and flame etc.

Storage and Stability

Avoid direct sun light, avoid high temperature, avoid high humidity, and avoid dust.

Legal Information

Product of LG Electronics, R&D use only

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinglei Ping et al.
ACS nano, 10(9), 8700-8704 (2016-08-18)
Scalable production of all-electronic DNA biosensors with high sensitivity and selectivity is a critical enabling step for research and applications associated with detection of DNA hybridization. We have developed a scalable and very reproducible (>90% yield) fabrication process for label-free
Omid Akhavan
Journal of materials chemistry. B, 4(19), 3169-3190 (2016-05-21)
Although graphene/stem cell-based tissue engineering has recently emerged and has promisingly and progressively been utilized for developing one of the most effective regenerative nanomedicines, it suffers from low differentiation efficiency, low hybridization after transplantation and lack of appropriate scaffolds required
Bing Deng et al.
Advanced materials (Deerfield Beach, Fla.), 31(9), e1800996-e1800996 (2018-10-03)
Chemical vapor deposition (CVD) is considered to be an efficient method for fabricating large-area and high-quality graphene films due to its excellent controllability and scalability. Great efforts have been made to control the growth of graphene to achieve large domain

Articles

Review on 1D vdWHs: Discusses materials, synthesis, optoelectronic applications, challenges, and future perspectives for 1D vdWH-based devices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service