Skip to Content
Merck
All Photos(2)

Key Documents

919942

Sigma-Aldrich

NanoFabTx reagent

PEGPLA-L-M

Synonym(s):

NanoFabTx

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$703.00

$703.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$703.00

About This Item

Empirical Formula (Hill Notation):
C6H10O2·C2H4O)x·CH4O
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

$703.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Quality Level

application(s)

advanced drug delivery

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

NanoFabTx reagent, PEGPLA-L-M is a component of our NanoFabTx PEG-PLA drug formulation screening kit (920436), which includes additional reagents and protocols for nanoprecipitation and microfluidics to synthesize 100-200 nm nanoparticles.NanoFabTx reagent, PEGPLA-L-M is a pegylated polylactic acid (PLA) polymer. PLA is biocompatible and biodegradable and has been approved by the FDA for biomedical and pharmaceutical applications.

Application

NanoFabTx reagent, PEGPLA-L-M can be used as a refill for the component in our NanoFabTx PEG-PLA drug formulation screening kit (920436).

Legal Information

NANOFABTX is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service