Skip to Content
Merck

Skip To

919829

Poly(styrene-(phenyl-13C6))

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
[-CH(13C6H5)CH2-]n
UNSPSC Code:
12162002
Isotopic purity:
≥99 atom % 13C
Mass shift:
M+6 (per repeating unit)
Form:
solid
Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service


isotopic purity

≥99 atom % 13C

form

solid

mass shift

M+6 (per repeating unit)


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
604453604445600067
description

-

description

99 atom % 13C

description

99 atom % 13C

description

99 atom % 13C

isotopic purity

≥99 atom % 13C

isotopic purity

99 atom % 13C

isotopic purity

99 atom % 13C

isotopic purity

99 atom % 13C

mass shift

M+6 (per repeating unit)

mass shift

M+1 (per repeating unit)

mass shift

M+1 (per repeating unit)

mass shift

M+1 (per repeating unit)

form

solid

form

solid

form

solid

form

solid


Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters