Skip to Content
Merck
All Photos(1)

Key Documents

91917

Sigma-Aldrich

5,5,5-Trifluoro-DL-leucine

≥98.0% (sum of isomers, HPLC)

Synonym(s):

(±)-2-Amino-4-(trifluoromethyl)pentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10F3NO2
CAS Number:
Molecular Weight:
185.14
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥98.0% (sum of isomers, HPLC)

form

crystals

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

CC(CC(N)C(O)=O)C(F)(F)F

InChI

1S/C6H10F3NO2/c1-3(6(7,8)9)2-4(10)5(11)12/h3-4H,2,10H2,1H3,(H,11,12)

InChI key

XFGVJLGVINCWDP-UHFFFAOYSA-N

General description

5,5,5-Trifluoro-DL-leucine, also known as trifluoroleucine, is an analog of L-leucine amino acid, often used to synthesize highly fluorinated peptides.[1]

Application

5,5,5-trifluoroleucine can be incorporated into the coiled-coil peptides to increase their thermal stability.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiro Oba et al.
Bioscience, biotechnology, and biochemistry, 70(7), 1776-1779 (2006-07-25)
We characterized a trifluoroleucine-resistant mutant of Saccharomyces cerevisiae, TFL20, that has a mutation in the LEU4 gene. We monitored the concentration of extracellular i-AmOH and intracellular amino acids, and compared the ratios of gene expression in TFL20 with the wild-type
A Niemz et al.
Journal of the American Chemical Society, 123(30), 7407-7413 (2001-07-27)
We have investigated the effect of trifluoroleucine substitution on the membrane-binding and tetramerization behavior of melittin. Analogues were synthesized in which Leu 9, Leu 13, and all four intrinsic leucine residues of melittin were replaced by 5,5,5-trifluoroleucine. Both the mono-
B Bendoni et al.
FEMS microbiology letters, 180(2), 229-233 (1999-11-11)
The resistance to 5,5,5-trifluoro-DL-leucine, encoded by the dominant allele LEU4-1, was used as a selectable marker to transform laboratory and natural Saccharomyces cerevisiae strains by the lithium acetate procedure. Results of transformation of S. cerevisiae laboratory and wine natural strains
E Casalone et al.
Research in microbiology, 148(7), 613-623 (1998-10-10)
Eighteen mutants resistant to 5',5',5'-trifluoroleucine (TFL), a leucine analog, were isolated in Saccharomyces cerevisiae strains YNN281 and YNN282. The mutants were characterized genetically and clustered in two groups, one comprising all the dominant (TFL1) and the other one all the
Tatyana Panchenko et al.
Biotechnology and bioengineering, 94(5), 921-930 (2006-03-21)
Varied levels of fluorinated amino acid have been introduced biosynthetically to test the functional limits of global substitution on enzymatic activity and stability. Replacement of all the leucine (LEU) residues in the enzyme chloramphenicol acetyltransferase (CAT) with the analog, 5',5',5'-trifluoroleucine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service