Skip to Content
Merck
All Photos(1)

Key Documents

918776

Sigma-Aldrich

PIM-py

Synonym(s):

2,3,5,6-Tetrafluoro-4-pyridinecarbonitrile-3,3,3′,3′-tetramethyl-1,1′-spirobisin dane-5,5′,6,6′-tetrol co-polymer, Polymers of intrinsic microporosity

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 G
$910.00

$910.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2.5 G
$910.00

About This Item

Linear Formula:
(C27H20N2O4)n
UNSPSC Code:
12162002
NACRES:
NA.23

$910.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Application

Polymers of intrinsic microporosity (PIMs) have generated significant interest for membrane separations and other applications since their initial discovery in 2004. PIMs are designed to have highly rigid and contorted backbone structures that cannot pack efficiently and thus exhibit microporosity. PIMs find applications in industry as they are solution processable, have a high permeability, and exhibit significant gas pair selectivities as membranes. [1][2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymers of Intrinsic Microporosity.
McKeown N B, et al.
ISRN Materials Science, 7-7 (2012)
Understanding the Topology of the Polymer of Intrinsic Microporosity PIM-1: Cyclics, Tadpoles, and Network Structures and Their Impact on Membrane Performance.
Foster A B, et al.
Macromolecules, 53(2), 569-583 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service