Skip to Content
Merck
All Photos(1)

Key Documents

91681

Sigma-Aldrich

N-(5-Hydroxypentyl)trifluoroacetamide

≥98.0% (GC)

Synonym(s):

5-(Trifluoroacetamido)-1-pentanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12F3NO2
CAS Number:
Molecular Weight:
199.17
Beilstein:
6644762
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

form

liquid

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.420

density

1.248 g/mL at 20 °C (lit.)

SMILES string

OCCCCCNC(=O)C(F)(F)F

InChI

1S/C7H12F3NO2/c8-7(9,10)6(13)11-4-2-1-3-5-12/h12H,1-5H2,(H,11,13)

InChI key

SCHGQSPQAJAJRI-UHFFFAOYSA-N

Other Notes

Selective mono-O-alkylation is accomplished by N,O-dianion formation prior to alkylation; otherwise, preferential N-alkylation takes place[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Hirschmann et al.
Tetrahedron Letters, 36, 2373-2373 (1995)
R. Hirschmann et al.
Journal of the American Chemical Society, 115, 12550-12550 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service