Skip to Content
Merck
All Photos(1)

Key Documents

916773

Sigma-Aldrich

Tip caps for cartridges

one size, blue, 50/box

Synonym(s):

3d bioprinting consumables

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$30.70

$30.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$30.70

About This Item

UNSPSC Code:
41121800
NACRES:
NA.23

$30.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

End caps (50/box), blue
Suitable for 3mL cartridge

Quality Level

Application

Tip caps have a large knurled gripping surface that simplifies attachment, and a vent that prevents air from being introduced into the syringe barrel during installation. The gripping action of the tip cap is designed to maximize the seal and yet be easilyremoved by the user. End caps and tip caps provide an airtight seal that allows you to prefill cartridges or seal partially used syringes between shifts.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service