Skip to Content
Merck
All Photos(2)

Documents

915378

Sigma-Aldrich

AdBrettPhos Pd G6 Br

Synonym(s):

AdBrettPhos OAC precatalyst Br

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H65BrF3O2PPd
Molecular Weight:
972.34
UNSPSC Code:
12352100
NACRES:
NA.22

form

powder or crystals

Quality Level

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

Application

AdBrettPhos Pd G6 Br is a G6 Buchwald precatalyst. The G6 precatalysts are air-stable, palladium-based oxidative addition complexes (OACs) that offer an alternative to the previously developed classes of palladacycle precatalysts. Unlike previous generations, the bulkiest biarylphosphine ligand precatalysts are readily prepared. These precatalysts have been shown to be effective for many reactions, including C-N, C-O, and C-F cross couplings.

Learn more about G6 Buchwald precatalysts

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Buchwald G6 precatalysts are oxidative addition complexes that are thermally, air, and moisture stable. The catalyst activation doesn’t require a base and generates innocuous byproducts.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service