Skip to Content
Merck
All Photos(2)

Key Documents

915041

Sigma-Aldrich

Platinum nanospheres

citrate functionalized, 50 nm diameter, 0.05 mg/mL in 2 mM aqueous sodium citrate buffer

Synonym(s):

Platinum nanoparticles, PtNP

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$493.00

$493.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$493.00

About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.23

$493.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Quality Level

form

nanospheres

storage temp.

2-8°C

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Platinum nanoparticles have many applications ranging from the automotive, chemical, and biomedical industries.[1][2][3]

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deborah Pedone et al.
Chemical Society reviews, 46(16), 4951-4975 (2017-07-12)
Oxidative stress-dependent inflammatory diseases represent a major concern for the population's health worldwide. Biocompatible nanomaterials with enzymatic properties could play a crucial role in the treatment of such pathologies. In this respect, platinum nanoparticles (PtNPs) are promising candidates, showing remarkable
Recent Advances in FePt Nanoparticles for Biomedicine.
Shi Y, et al.
Journal of Nanomaterials, 2015, 1-13 (2015)
Pt-based composite nanoparticles for magnetic, catalytic, and biomedical applications.
Liu Y, et al.
Journal of Materials Chemistry, 21, 12579-12587 (2011)

Articles

Research on noble bimetallic nanoparticles for glucose biosensor detection.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service