Skip to Content
Merck
All Photos(1)

Key Documents

914983

Sigma-Aldrich

ThT-NE

≥95%

Synonym(s):

2-(4-(Diethylamino)phenyl)-3,6-dimethylbenzo[d]thiazol-3-ium chloride, Fluorogenic viral RNA probe, G-Quadruplex Probe, Viral RNA light-up probe

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$563.00

$563.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$563.00

About This Item

Empirical Formula (Hill Notation):
C19H23ClN2S
CAS Number:
Molecular Weight:
346.92
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

$563.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

storage temp.

−20°C

SMILES string

[s]1c2c([n+](c1c3ccc(cc3)N(CC)CC)C)ccc(c2)C

InChI key

WPIXUKQQLRQGGY-UHFFFAOYSA-N

Application

ThT-NE is a benzothiazole-based G-quadruplex (G4)-targeted fluorogenic probe developed by Zhou Nie and coworkers for the visualization of viral RNA in living host cells, including staining, real-time, and live-cell tracking applications.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service