913324
2- Methyloxyethylammonium iodide
Synonym(s):
2- Methyloxyethylammonium iodide, 2-MeO-EAI, Ethanamine, 2-methoxy-, hydriodide (1:1), Greatcell Solar®
Select a Size
Select a Size
About This Item
Recommended Products
form
powder
Quality Level
color
white
SMILES string
I.NCCOC
InChI
1S/C3H9NO.HI/c1-5-3-2-4;/h2-4H2,1H3;1H
InChI key
SCDXFIYEBSXILC-UHFFFAOYSA-N
Related Categories
Application
Legal Information
Greatcell Solar is a registered trademark of Greatcell Solar Materials Pty Ltd.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service