Skip to Content
Merck
All Photos(2)

Key Documents

912727

Sigma-Aldrich

(2R,3R)-iPr-BIDIME

Synonym(s):

(2R,3R)-3-(Tert-butyl)-4-(2,6-dimethoxyphenyl)-2-isopropyl-2,3-dihydrobenzo[d][1,3]oxaphosphole

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$407.00

$407.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$407.00

About This Item

Empirical Formula (Hill Notation):
C22H29O3P
CAS Number:
Molecular Weight:
372.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

$407.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

form

powder and chunks

Quality Level

optical purity

ee: ≥99% (HPLC)

reaction suitability

reagent type: ligand

functional group

phosphine

SMILES string

P1([C@@H](Oc2c1c(ccc2)c3c(cccc3OC)OC)C(C)C)C(C)(C)C

InChI

1S/C22H29O3P/c1-14(2)21-25-18-13-8-10-15(20(18)26(21)22(3,4)5)19-16(23-6)11-9-12-17(19)24-7/h8-14,21H,1-7H3/t21-,26?/m1/s1

InChI key

AVUPKIFOTAYCAC-OSMGYRLQSA-N

Related Categories

Application

(2R,3R)-iPr-BIDIME is a P-chiral monophosphorus ligand used for the transition metal-catalyzed asymmetric Suzuki-Miyaura and hydroboration reactions.

Legal Information

Sold in collaboration with Zejun Pharmaceuticals

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guangqing Xu et al.
Journal of the American Chemical Society, 136(2), 570-573 (2013-10-24)
Efficient asymmetric Suzuki-Miyaura coupling reactions are employed for the first time in total syntheses of chiral biaryl natural products korupensamine A and B in combination with an effective diastereoselective hydrogenation, allowing ultimately a concise and stereoselective synthesis of michellamine B.
Naifu Hu et al.
Journal of the American Chemical Society, 137(21), 6746-6749 (2015-05-06)
The rhodium-catalyzed asymmetric hydroboration of α-arylenamides with BI-DIME as the chiral ligand and (Bpin)2 as the reagent yields for the first time a series of α-amino tertiary boronic esters in good yields and excellent enantioselectivities (up to 99% ee).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service