Skip to Content
Merck
All Photos(3)

Key Documents

912301

Sigma-Aldrich

di-n-butylammonium tetramethylammonium lead(II) hexadecaiodide

greener alternative

n = 5

Synonym(s):

(BA)2(MA)4Pb5I16, 2D perovskite, Bis(butylammonium) tetrakis(methylammonium) hexadecaiodopentaplumbate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$532.00

$532.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$532.00

About This Item

Empirical Formula (Hill Notation):
C12H48N6Pb5I16
Molecular Weight:
3343.02
UNSPSC Code:
12352101
NACRES:
NA.23

$532.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

[NH3+]C.[NH3+]C.CCCC[NH3+].CCCC[NH3+].[NH3+]C.[NH3+]C

InChI

1S/2C4H11N.4CH5N/c2*1-2-3-4-5;4*1-2/h2*2-5H2,1H3;4*2H2,1H3/p+6

InChI key

JHERJNURAJDMBS-UHFFFAOYSA-T

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for "Energy efficiency".

Application

2D perovskites have been used as the active material in may applications such as light emmiting diodes,[1] phototransistors,[2] and solar cells.[3] Unlike 3D perovskites, these layered materials give higher moisture stability and longer device lifetimes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Duyen H Cao et al.
Journal of the American Chemical Society, 137(24), 7843-7850 (2015-05-29)
We report on the fabrication and properties of the semiconducting 2D (CH3(CH2)3NH3)2(CH3NH3)(n-1)Pb(n)I(3n+1) (n = 1, 2, 3, and 4) perovskite thin films. The band gaps of the series decrease with increasing n values, from 2.24 eV (CH3(CH2)3NH3)2PbI4 (n = 1)
Mingjian Yuan et al.
Nature nanotechnology, 11(10), 872-877 (2016-06-28)
Organometal halide perovskites exhibit large bulk crystal domain sizes, rare traps, excellent mobilities and carriers that are free at room temperature-properties that support their excellent performance in charge-separating devices. In devices that rely on the forward injection of electrons and
Yuchuan Shao et al.
Nano letters, 17(12), 7330-7338 (2017-11-08)
Recently, two-dimensional (2D) organic-inorganic perovskites emerged as an alternative material for their three-dimensional (3D) counterparts in photovoltaic applications with improved moisture resistance. Here, we report a stable, high-gain phototransistor consisting of a monolayer graphene on hexagonal boron nitride (hBN) covered

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service