Skip to Content
Merck
All Photos(2)

Key Documents

910791

Sigma-Aldrich

Poly(ethylene glycol)-block-polyethyleneimine

PEG Mn 750, PEI Mn 15k

Synonym(s):

Linear poly(ethylene glycol)-block-polyethylenimine, PEG-b-PEI, PEG-PEI

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$424.00

$424.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$424.00

About This Item

Linear Formula:
CH3(C2H4O)m(C2H5N)nOH
UNSPSC Code:
12162002
NACRES:
NA.23

$424.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder or solid

mol wt

PEG Mn 750
PEG Mw 750 Da
PEI Mn 15k
PEI Mw 10,000 Da

color

white to pale yellow

storage temp.

2-8°C

Application

Gene therapy using polymeric nonviral vectors promises considerable advances in the treatment of many genetic and acquired diseases due to the carrier safety and low immunogenicity profiles. Poly(ethyleneimine) is one of the most commonly used cationic polymers for gene delivery applications. Copolymers of PEI and hydrophilic poly(ethylene glycol) have been designed to provide a stealth-like shield around polymer/DNA complexes, reducing nonspecific interactions, inhibiting activation of the reticuloendothelial system, and prolonging the half-life of the complexes in the blood.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pallab Banerjee et al.
Bioconjugate chemistry, 17(1), 125-131 (2006-01-19)
A block copolymer of a hyperbranched poly(ethylene glycol)-like core and linear polyethylenimine (HBP) was synthesized by a facile synthetic route that included (1) a single-step cationic copolymerization of diepoxy and polyhydroxyl monomers, (2) derivatization of hydroxyl groups of the core
Dagmar Fischer et al.
Drug metabolism and disposition: the biological fate of chemicals, 32(9), 983-992 (2004-08-21)
The in vivo body distribution and the pharmacokinetics of a 20mer double-stranded nuclear factor kappaB decoy oligodeoxynucleotide (ODN) complexed with 25-kDa poly(ethylene imine) (PEI), low molecular weight 2.7-kDa PEI, and PEGylated PEI [bPEI(25k)-glPEG(550)(50)] after intravenous injection were studied in BALB/c
B Brissault et al.
Biomacromolecules, 7(10), 2863-2870 (2006-10-10)
The study of ethyloxazoline/methyloxazoline (EtOXZ/MeOXZ) copolymerization, initiated by methyl tosylate (MeOTs), showed that (i) incorporation of MeOXZ units into random copolymer becomes effective over DP = 100 and (ii) propagation process proceeds with negligible transfer to monomer up to a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service