Skip to Content
Merck
All Photos(1)

Documents

909033

Sigma-Aldrich

2,7-Dibromo-9-mesityl-10-methylacridinium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H20BBr2F4N
Molecular Weight:
557.02
UNSPSC Code:
12352101
NACRES:
NA.22

form

powder or crystals

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

Related Categories

Application

Robust acridinium-based photocatalyst for use as an alternative to transition-metal-based photocatalysts with higher chemical stability and attenuated redox potential.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Nicewicz lab is focused on the discovery of new and powerful reaction methodologies that proceed via the intermediacy of highly reactive cation radical species. Included in these transformations are anti-Markovnikov selective additions of amines, alcohols, carboxylic acids, amides and mineral acids to alkenes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service