Skip to Content
Merck
All Photos(2)

Key Documents

908487

Sigma-Aldrich

N-(tert-butyl)-N-((ethoxycarbonothioyl)thio)-2,3,4,5,6-pentafluorobenzamide

≥95%

Synonym(s):

Alexanian xanthylation reagent

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$221.00

$221.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$221.00

About This Item

Empirical Formula (Hill Notation):
C14H14F5NO2S2
Molecular Weight:
387.39
UNSPSC Code:
12352111
NACRES:
NA.22

$221.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

≥95%

form

powder or crystals

mp

46-48 °C

functional group

amine
fluoro

storage temp.

−20°C

Application

As reported by the Alexanian laboratory, this reagent is used for site-selective, intermolecular decarboxylative xanthylation leading to the rapid diversification of carboxylic acids. Once installed, the xanthate group provides direct access to diverse product analogues via several aliphatic C-H transformations, including halogenation, deuteration, vinylation, hydroxylation. Product is best stored in foil-wrapped vials in the freezer when not in use; however, it can be weighed out on the bench without risk of decomposition.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service