Skip to Content
Merck
All Photos(3)

Key Documents

906808

Sigma-Aldrich

Water-soluble TPO based nanoparticle photoinitiator

contains ionic surfactant

Synonym(s):

Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide, TPO

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$193.00
5 G
$764.00

$193.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$193.00
5 G
$764.00

About This Item

MDL number:
UNSPSC Code:
12352005
NACRES:
NA.25

$193.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

form

powder or solid

color

white to off-white

SMILES string

O=P(C1=CC=CC=C1)(C(C2=C(C)C=C(C)C=C2C)=O)C3=CC=CC=C3

InChI

1S/C22H21O2P/c1-16-14-17(2)21(18(3)15-16)22(23)25(24,19-10-6-4-7-11-19)20-12-8-5-9-13-20/h4-15H,1-3H3

InChI key

VFHVQBAGLAREND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The water-dispersible photoinitiator nanoparticle contains 10% (w/w) of the type I photoinitiator diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide (TPO). TPO is highly efficient but water-insoluble. This water-soluble TPO based photoinitiator enables rapid 3D printing of structures prepared in aqueous solutions while bringing environmental advantages by using low-energy curing systems and avoiding the need for solvents.

The extinction coefficient of the new water-dispersible nanoparticles of TPO is more than 300 times larger than the best and most used commercially available water-soluble photoinitiator, Irgacure 2959. The TPO nanoparticles absorb significantly in the range from 385 to 420 nm, making them suitable for use in commercially available, low-cost, light-emitting diode-based 3D printers and UV-curing devices.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3D printing of responsive hydrogels for drug-delivery systems
Magdassi S. et al.
Journal of 3D printing in medicine, 1(4), 219-229 (2017)
Amol A Pawar et al.
Science advances, 2(4), e1501381-e1501381 (2016-04-07)
In the absence of water-soluble photoinitiators with high absorbance in the ultraviolet (UV)-visible range, rapid three-dimensional (3D) printing of hydrogels for tissue engineering is challenging. A new approach enabling rapid 3D printing of hydrogels in aqueous solutions is presented on

Articles

Water-dispersible photoinitiator nanoparticles enable novel formulations for 3D bioprinting, tissue engineering, and device manufacturing.

Related Content

Tissue engineering fabricates tissues cultures from scaffolds, living cells, and biologically active molecules by simulating the microenvironment of the body to repair or replace damaged tissue.

Questions

  1. What is the solubility of "906808 Water-soluble TPO based nanoparticle photoinitiator

    1 answer
    1. This product is soluble in water. A rate has not been established.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service