Skip to Content
Merck
All Photos(1)

Key Documents

905089

Sigma-Aldrich

In Vitro Protein Expression (iPE) Kit II

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$1,110.00

$1,110.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 KIT
$1,110.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.12

$1,110.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

technique(s)

bio NMR: suitable
protein expression: suitable

Quality Level

storage temp.

−70°C

Related Categories

Application

iPE-Quick Kit (prod.no. 767824) is available and intended for the confirmation of target protein expression utilizing E. Coli extract before the use of this iPE kit.
This is a protein synthesis system that utilizes E. coli cell extract. It allows easy and efficient protein expression by adding circular or linear DNA as the template DNA, which enables transcription of mRNA with T7 RNA polymerase.
This kit has been developed under license from RIKEN, incorporating their proprietary, advanced cell-free protein synthesizing technology into a kit dedicated to stable isotope labeling.
This kit is not intended to be used for disulfide-containing proteins. For disulfide-containing proteins, refer to prod.no. 797006.

Packaging

For information on pricing and availability, please contact Stable Isotopes Customer Service.
Stable isotope labeled amino acid mixtures are not included in this kit. Please use separately-sold stable isotope labeled amino acids mixtures (prod. no. 767972, 767964, 771031).

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Yabuki et al.
Journal of structural and functional genomics, 8(4), 173-191 (2008-01-02)
A two-step PCR method has been developed for the robust, high-throughput production of linear templates ready for cell-free protein synthesis. The construct made from the cDNA expresses a target protein region with N- and/or C-terminal tags. The procedure consists only
Takayoshi Matsuda et al.
Journal of biomolecular NMR, 37(3), 225-229 (2007-01-24)
Cell-free protein synthesis is suitable for stable-isotope labeling of proteins for NMR analysis. The Escherichia coli cell-free system containing potassium acetate for efficient translation (KOAc system) is usually used for stable-isotope labeling, although it is less productive than other systems.
Eiko Seki et al.
Analytical biochemistry, 377(2), 156-161 (2008-04-01)
Cell-free protein synthesis has become one of the standard methods for protein expression. One of the major advantages of this method is that PCR-amplified linear DNA fragments can be directly used as templates for protein synthesis. The productivity of cell-free
William C Yang et al.
Biotechnology progress, 28(2), 413-420 (2012-01-26)
Escherichia coli cell-free protein synthesis (CFPS) uses E. coli extracts to make active proteins in vitro. The basic CFPS reaction mixture is comprised of four main reagent components: (1) energy source and CFPS chemicals, (2) DNA encoding the protein of
Jun Yokoyama et al.
Analytical biochemistry, 411(2), 223-229 (2011-01-25)
During recent years, the targets of protein structure analysis using nuclear magnetic resonance spectroscopy have become larger and more complicated. As a result, a complete and precise stable isotope labeling technique has been desired. A cell-free protein synthesis system is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service