Skip to Content
Merck
All Photos(1)

Key Documents

904163

Sigma-Aldrich

Glycochenodeoxycholic-2,2,4,4-d4 acid solution

100 μg/mL in methanol, ≥98 atom % D, ≥97% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$508.00

$508.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 ML
$508.00

About This Item

Empirical Formula (Hill Notation):
C26D4H39NO5
Molecular Weight:
453.65
UNSPSC Code:
12352106
NACRES:
NA.24

$508.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

≥98 atom % D

Quality Level

Assay

≥97% (CP)

form

solution

concentration

100 μg/mL in methanol

technique(s)

mass spectrometry (MS): suitable

mass shift

M+4

storage temp.

−20°C

SMILES string

O[C@H](C1)C(C2[C@]([C@@H]([C@H](C)CCC(NCC(O)=O)=O)CC2)(C)CC3)C3[C@]4(C)[C@@]1([H])C([2H])([2H])[C@H](O)C([2H])([2H])C4

InChI

1S/C26H43NO5/c1-15(4-7-22(30)27-14-23(31)32)18-5-6-19-24-20(9-11-26(18,19)3)25(2)10-8-17(28)12-16(25)13-21(24)29/h15-21,24,28-29H,4-14H2,1-3H3,(H,27,30)(H,31,32)/t15-,16+,17-,18-,19?,20?,21-,24?,25+,26-/m1/s1/i8D2,12D2

InChI key

GHCZAUBVMUEKKP-KWFHFAJKSA-N

Related Categories

Packaging

1 mL in ampule
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service