Skip to Content
Merck
All Photos(1)

Documents

902268

Sigma-Aldrich

seMpai

Synonym(s):

DP3-Na, TokeOni analogue, water-soluble NIR emission luciferin analogue, 2-[(1E,3E)-6-[3-(dimethylamino)pyridine]-1,3-butadien-1-yl]-4,5-dihydro-(4S)-4-thiazolecarboxylic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16N3NaO2S
Molecular Weight:
325.36
UNSPSC Code:
12352106
NACRES:
NA.23

description

Purity: mixture of D (80% min.) and L (20% max.) forms

form

powder

color

black to yellow-brown

storage temp.

−20°C

General description

2-[(1E,3E)-6-[3-(dimethylamino)pyridine]-1,3-butadien-1-yl]-4,5-dihydro-(4S)-4-thiazolecarboxylic acid sodium salt (DP3-Na) is a new NIR emission luciferin analog having luminescence peak at 670 ~ 680 nm with a peak range in the near-infrared (NIR) region with the enhanced solubility in water.
Luciferin is a generic term for the light-emitting compound found in organisms that generate bioluminescence.

Application

Applications:
  • Bioimaging
  • In vivo imaging

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantum yield improvement of red-light-emitting firefly luciferin analogues for in vivo bioluminescence imaging
Kiyama M, et al.
Tetrahedron, 74(6), 652- 660 (2018)
High Sensitivity In Vivo Imaging of Cancer Metastasis Using a Near-Infrared Luciferin Analogue seMpai
Nakayama J, et al.
International Journal of Molecular Sciences, 21(21), 7896-7896 (2020)
Visualization of activity-regulated BDNF expression in the living mouse brain using non-invasive near-infrared bioluminescence imaging.
Fukuchi M, et al.
Molecular Brain, 13(1), 122-122 (2020)
Synthesis and Luminescence Properties of Near-Infrared N-Heterocyclic Luciferin Analogues for In Vivo Optical Imaging.
Saito R, et al.
Bulletin of the Chemical Society of Japan, 92(3), 182-8585 (2018)

Articles

seMpai substrate is suitable for near-infrared BLI in biological experiments, offering high solubility for extended bioluminescent applications.

Bioluminescence imaging (BLI) systems allows for high-sensitive and noninvasive monitoring of cell proliferation, activity of signaling pathways and protein-protein interactions in living tissues.

Graphene nanoribbons (GNRs) are quasi-one-dimensional narrow strips of graphene comprised of sp2-hybridized carbon atoms arranged into hexagonal honeycomb lattice configurations.

Related Content

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service