Skip to Content
Merck
All Photos(1)

Key Documents

901252

Sigma-Aldrich

2,6-Dimethylaniline hydrochloride

≥98%

Synonym(s):

2,6-Dimethylbenzenamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$55.50
5 G
$151.00

$55.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$55.50
5 G
$151.00

About This Item

Empirical Formula (Hill Notation):
C8H12ClN
CAS Number:
Molecular Weight:
157.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

$55.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98%

form

powder or crystals

mp

275 °C

SMILES string

[Cl-].[N+H3]c1c(cccc1C)C

InChI

1S/C8H11N.ClH/c1-6-4-3-5-7(2)8(6)9;/h3-5H,9H2,1-2H3;1H

InChI key

QNWMFJDRMZWZNN-UHFFFAOYSA-N

Application

2,6-Dimethylaniline hydrochloride may be used in the preparation of 2,6-dimethylphenyl isocyanate via phosgenation. This product was previously listed as CDS000447.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Urea dissociation. Measure of steric hindrance in secondary amines.
Stowell JC and Padegimas SJ.
The Journal of Organic Chemistry, 39 (16), 2448-2449 (1974)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service