Skip to Content
Merck
All Photos(1)

Key Documents

901212

Sigma-Aldrich

5-Methyl-5-propargyloxycarbonyl-1,3-dioxane-2-one

≥98.0%

Synonym(s):

Propargyl 5-methyl-2-oxo-1,3-dioxane-5-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O5
CAS Number:
Molecular Weight:
198.17
UNSPSC Code:
12162002

Assay

≥98.0%

form

powder or chunks

color

colorless to pale yellow

storage temp.

−20°C

General description

5-Methyl-5-propargyloxycarbonyl-1,3-dioxane-2-one is an alkyne-functionalized, cyclic carbonate monomer that can be polymerized or copolymerized to yield alkyne-functionalized polycarbonates. The incorporation of alkynes allows for rapid post-polymerization modification through reactions, such as copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) or thiol-yne click reactions. These reactions are highly preferable for functionalization due their high efficiency and the ability to easily install the corresponding functionality (e.g., azides or thiols) onto the biomolecule or small molecule of interest.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of ferrocene-functionalized monomers for biodegradable polymer formation.
Upton BM, et al.
Inorganic Chemistry, 1, 271-271 (2014)
Russell C Pratt et al.
Chemical communications (Cambridge, England), (1), 114-116 (2008-04-11)
Cyclic carbonate monomers based on a single biocompatible scaffold allow for incorporation of a wide range of functional groups into macromolecules via ring-opening polymerization.
Organocatalytic synthesis and post-polymerization functionalization of propargyl-functional poly(carbonate)s.
Tempelaar S, et al.
Polym. Chem., 4, 174-174 (2013)
Daniel P Sanders et al.
Journal of the American Chemical Society, 132(42), 14724-14726 (2010-10-05)
An improved two-step synthetic route to functionalized cyclic carbonate monomers that features a novel cyclic carbonate intermediate with an active pentafluorophenyl ester group (MTC-OPhF(5)) has been developed. The versatile pentafluorophenyl ester intermediate can be synthesized on the gram to kilogram

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service