Skip to Content
Merck
All Photos(1)

Key Documents

901016

Sigma-Aldrich

1-Propyl-2,3-dimethyl-imidazolium iodide Solarpur®

Synonym(s):

1,2-Dimethyl-3-propylimidazolium iodide, DMPII, PMMIM I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15N2I
CAS Number:
Molecular Weight:
266.12
UNSPSC Code:
12352200
NACRES:
NA.23

Assay

≥99.0% (HPLC)

form

powder or crystals

availability

available only in USA

InChI

1S/C8H15N2.HI/c1-4-5-10-7-6-9(3)8(10)2;/h6-7H,4-5H2,1-3H3;1H/q+1;/p-1

InChI key

ISHFYECQSXFODS-UHFFFAOYSA-M

Application

Solarpur® electrolyte components for DSSC applications meet the highest purity standards regarding water and other impurities required for this technology.

Legal Information

Solarpur® is a trademark of EMD Performance Materials Corp.
Solarpur is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service