900847
4-(Fluorosulfonyl)-3-((trimethylsilyl)ethynyl)benzoic acid
≥95%
Synonym(s):
Carboxyl sulfonyl fluoride TMS-alkyne, Probe building block
About This Item
Assay
≥95%
form
powder or crystals
storage temp.
−20°C
SMILES string
OC(C1=CC(C#C[Si](C)(C)C)=C(S(F)(=O)=O)C=C1)=O
Application
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
In partnership with Pfizer chemists, we have compiled a collection of trifunctional building blocks to enable development of probes.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service