Skip to Content
Merck
All Photos(1)

Key Documents

900769

Sigma-Aldrich

Silica

mesoporous MCM-48, <15 μm particle size, pore size 3 nm, amine functionalized

Synonym(s):

Amine functionalized MCM-48, Amine functionalized mesoporous silica, Mesoporous silica

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$1,480.00

$1,480.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$1,480.00

About This Item

UNSPSC Code:
12352309
NACRES:
NA.23

$1,480.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

powder

particle size

<15 μm

pore size

3 nm pore size

functional group

amine

Looking for similar products? Visit Product Comparison Guide

Application

The unique physical properties such as high surface area and ordered pore structure of mesoporous silica combined with the selectivity of amine and thiol functional groups make them useful for diverse applications. For example amine and thiol modified mesoporous silica finds applications in CO2 sequestration, heavy metal (such as Pb, Fe, Au etc.) removal from waste water or industrial waste streams.[1][2] Also, the ability of the amine and thiol modified mesoporous silica to bind the metallic nanoparticles such as Pt or Pd render them useful as novel catalytic support.[3] The functionalization of mesoporous silica with amine and thiol also makes them useful for entrapping biological entities and for investigations involving the study of the effect of quantum confinements of molecules/solvent within the ordered mesopores.[4][5]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hydrogen Bonding of Water Confined in Mesoporous Silica MCM-41 and SBA-15 Studied by 1H Solid-State NMR.
Grunberg B, et al.
Chemistry?A European Journal , 10(22), 5689-5696 (2004)
Large pore bi-functionalised mesoporous silica for metal ion pollution treatment.
Burke AM, et al.
Journal of Hazardous Materials, 164(1), 229-234 (2009)
Synthesis of Gold Catalysts Supported on Mesoporous Silica Materials: Recent Developments.
Gutierrez LF, et al.
Catalysts, 1(1), 97-154 (2011)
Chenghong Lei et al.
Journal of the American Chemical Society, 124(38), 11242-11243 (2002-09-19)
The enzyme organophosphorus hydrolase (OPH) was spontaneously entrapped in carboxylethyl- or aminopropyl-functionalized mesoporous silica with rigid, uniform open-pore geometry (30 nm). This approach yielded larger amounts of protein loading and much higher specific activity of the enzyme when compared to
Amrit Kumar et al.
Angewandte Chemie (International ed. in English), 54(48), 14372-14377 (2015-10-07)
Sequestration of CO2, either from gas mixtures or directly from air (direct air capture, DAC), could mitigate carbon emissions. Here five materials are investigated for their ability to adsorb CO2 directly from air and other gas mixtures. The sorbents studied

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service