Skip to Content
Merck
All Photos(1)

Key Documents

900751

Sigma-Aldrich

Biotinylated-D-lysine TFA salt

Synonym(s):

(R)-2-Ammonio-6-(5-((3aR,4R,6aS)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)hexanoate 2,2,2-trifluoroacetate, BDL TFA salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$632.00

$632.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$632.00

About This Item

Empirical Formula (Hill Notation):
C16H28N4O4S · C2HF3O2
Molecular Weight:
486.51
UNSPSC Code:
12352209
NACRES:
NA.22

$632.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

form

powder or crystals

Quality Level

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

O=C(O)C(F)(F)F.[O-]C([C@@H](CCCCNC(CCCC[C@@H]1[C@@](NC(N2)=O)([H])[C@@]2([H])CS1)=O)[NH3+])=O

InChI

1S/C16H28N4O4S.C2HF3O2/c17-10(15(22)23)5-3-4-8-18-13(21)7-2-1-6-12-14-11(9-25-12)19-16(24)20-14;3-2(4,5)1(6)7/h10-12,14H,1-9,17H2,(H,18,21)(H,22,23)(H2,19,20,24);(H,6,7)/t10-,11-,12-,14-;/m1./s1

InChI key

ICPIMQLAGIDGFB-XSCHNBKYSA-N

Application

Biotinylated D-lysine (BDL) can be used to conjugate a biotin handle into bacterial peptidoglycan (PG) for various applications. For example, Suzanne Walker and coworkers reported that penicillin-binding protein PBP4 of Staphylococcus aureus could use BDL to biotinylate Lipid I and Lipid II, facilitating assays to measure infamously low-abundance pools of cellular lipid-linked PG precursors. BDL provides a tool for studying other aspects of PG assembly including antibacterial development efforts.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service