Skip to Content
Merck
All Photos(1)

Key Documents

900651

Sigma-Aldrich

3-Methacrylamidopropyltris(trimethylsiloxy)silane

>95%, stabilized with 4-Methoxyphenol, 50-500 ppm

Synonym(s):

N-[Tris(trimethylsiloxy)silylpropyl]methacrylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H39NO4Si4
CAS Number:
Molecular Weight:
421.83
UNSPSC Code:
12162002

Assay

>95%

form

liquid

concentration

50-500 ppm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

This monomer is commonly used in the formulation of research silicone hydrogel contact lenses and other research ophthalmic devices. Silicone monomers are preferred for the synthesis of research ophthalmic materials due to their high oxygen permeability.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The development of biomaterials for contact lens applications: effects of wear modality on materials design in Biomaterials and Regenerative Medicine in Ophthalmology.
Tighe, B.J. and Mann, A.
Biomaterials and Regenerative Medicine in Ophthalmology (Second Edition), 326-399 (2016)
Hydrophilic silicone hydrogels with interpenetrating network structure for extended delivery of ophthalmic drugs.
Wang J, et al.
Polymers For Advanced Technologies, 23 (9), 1258-1263 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service