Skip to Content
Merck
All Photos(1)

Key Documents

900579

Sigma-Aldrich

(S)-2-Benzylpiperidine

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$607.00

$607.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$607.00

About This Item

Empirical Formula (Hill Notation):
C12H17N
CAS Number:
Molecular Weight:
175.27
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

$607.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

Quality Level

Assay

≥95%

form

liquid

mp

46.2 °C

functional group

phenyl

storage temp.

2-8°C

SMILES string

N1[C@@H](CCCC1)Cc2ccccc2

InChI

1S/C12H17N/c1-2-6-11(7-3-1)10-12-8-4-5-9-13-12/h1-3,6-7,12-13H,4-5,8-10H2/t12-/m0/s1

InChI key

ITXCORRITGNIHP-LBPRGKRZSA-N

Application

Chiral building block developed using Liverpool ChiroChem-patented technology.

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service