Skip to Content
Merck
All Photos(1)

Key Documents

900544

Sigma-Aldrich

Rare earth doped upconversion nanoparticles

fluorescence λem 474 nm (main peak), 10 mg/mL in toluene

Synonym(s):

CANdot® Series X up blue, Luminescent nanoparticles, NaYF4:Yb, Tm/NaYF4, Upconversion nanocrystals

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$431.00

$431.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$431.00

About This Item

UNSPSC Code:
12352303
NACRES:
NA.23

$431.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

dispersion
nanoparticles

Quality Level

composition

NaYF4:Yb
Tm/NaYF4

concentration

10 mg/mL in toluene

particle size

17 nm

λmax

980 nm

fluorescence

λem 474 nm (main peak)
λem 474 nm±15 nm

functional group

oleic acid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Our rare earth doped upconversion nanoparticles possess inorganic fluoride host lattices and can be easily excited with near-infra-red (NIR: 980 nm) light source. These Cd-free nanoparticles exhibit characteristic emission peak and are well suited for applications as luminescent biological labels and as spectral converters for increasing solar cell efficiency cell.[1][2][3]

Legal Information

Fraunhofer CAN is a research division of the Fraunhofer IAP
CANdot is a registered trademark of Fraunhofer CAN

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

37.0 °F

Flash Point(C)

2.77 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced luminescence efficiency of aqueous dispersible NaYF4:Yb/Er nanoparticles and the effect of surface coating.
Ansari AA, et al.
Royal Society of Chemistry Advances, 6, 22074-22082 (2016)
Thorben Rinkel et al.
Angewandte Chemie (International ed. in English), 55(3), 1164-1167 (2015-12-04)
A new method is presented for preparing gram amounts of very small core/shell upconversion nanocrystals without additional codoping of the particles. First, ca. 5 nm β-NaYF4:Yb,Er core particles are formed by the reaction of sodium oleate, rare-earth oleate, and ammonium fluoride
Cytotoxic effects of upconversion nanoparticles in primary hippocampal cultures.
Vedunova MV, et al.
Royal Society of Chemistry Advances, 6, 33656-33665 (2016)

Questions

  1. What is the lifetime of these particles?

    1 answer
    1. This product does not contain an expiration date. It contains a recommended retest date of two years after the quality release date. A recommended retest date is the period of time during which the product is expected to remain within established stability specifications, provided that it has been stored under defined conditions. After the Retest Date, product samples should be examined to ensure that the product is still in compliance with the established specifications. For more information, you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service